Anti-FOXD3 Rabbit Recombinant Polyclonal Antibody
Catalog # 77843-863
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FOXD3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9UJU5
- Antigen Synonyms:AIS1|forkhead box D3|Forkhead box protein D3|foxd3|FOXD3_HUMAN|Genesis|HFH2|HNF3/FH transcription factor genesis|VAMAS2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Forkhead box protein D3 protein (1-140AA)
- Purification:>95%, Protein G purified
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human