Anti-AVPR1A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77851-709
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:AVPR1A
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P37288
- Antigen Synonyms:Antidiuretic hormone receptor 1a|Arginine vasopressin receptor 1A|AVPR 1|AVPR 1A|AVPR V1a|AVPR1|SCCL vasopressin subtype 1a receptor|V1 vascular vasopressin receptor V1a vasopressin receptor|V1aR|V1AR_HUMAN|Vascular/hepatic type arginine vasopressin receptor|Vascular/hepatic-type arginine vasopressin receptor|Vasopressin V1a receptor
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Vasopressin V1a receptor protein (1-52AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: AVPR1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human