Anti-CACNA1A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: CACNA1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77847-021
- 77847-022
- Antigen Symbol:
- CACNA1A
- CACNA1A
- Antigen Synonyms:
- CACNA1A| CACH4| CACN3| CACNL1A4| Voltage-dependent P/Q-type calcium channel subunit alpha-1A| Brain calcium channel I| BI| Calcium channel| L type| alpha-1 polypeptide isoform 4| Voltage-gated calcium channel subunit alpha Cav2.1
- CACNA1A| CACH4| CACN3| CACNL1A4| Voltage-dependent P/Q-type calcium channel subunit alpha-1A| Brain calcium channel I| BI| Calcium channel| L type| alpha-1 polypeptide isoform 4| Voltage-gated calcium channel subunit alpha Cav2.1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00555
- O00555
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human CACNA1A
- Synthetic peptide of Human CACNA1A
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together