Anti-AP1S1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-212
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:AP1S1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P61966
- Antigen Synonyms:Adapter related protein complex 1 sigma 1A subunit|Adapter-related protein complex 1 sigma-1A subunit|Adaptor protein complex AP-1 sigma-1A subunit|Adaptor protein complex AP1 sigma 1A subunit|AP-1 complex subunit sigma-1A|AP1 complex subunit sigma 1A|AP19|AP1S1|AP1S1_HUMAN|CLAPS1|Clathrin assembly protein complex 1 sigma 1A small chain|Clathrin assembly protein complex 1 sigma-1A small chain|Clathrin coat assembly protein AP19|Golgi adaptor HA1/AP1 adaptin sigma 1A subunit|Golgi adaptor HA1/AP1 adaptin sigma-1A subunit|HA1 19 kDa subunit|Sigma 1a subunit of AP-1 clathrin|Sigma 1a subunit of AP1 clathrin|Sigma adaptin 1A|Sigma-adaptin 1A|Sigma1A adaptin|Sigma1A-adaptin
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human AP-1 complex subunit sigma-1A protein (1-127AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: AP1S1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human