Anti-TMEFF2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: TMEFF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77847-757
- 77847-756
- Antigen Symbol:
- TMEFF2
- TMEFF2
- Antigen Synonyms:
- Cancer/testis antigen family 120 member 2|CT120.2|HPP 1|HPP1|Hyperplastic polyposis protein 1|TEFF 2|TEFF2|TEFF2_HUMAN|TENB 2|TENB2|TMEFF2|Tomoregulin|Tomoregulin-2|TPEF|TR 2|TR|TR-2|Transmembrane protein containing epidermal growth factor and follistatin domains|Transmembrane protein TENB2|Transmembrane protein with EGF like and two follistatin like domains 2|Transmembrane protein with EGF-like and two follistatin-like domains
- Cancer/testis antigen family 120 member 2|CT120.2|HPP 1|HPP1|Hyperplastic polyposis protein 1|TEFF 2|TEFF2|TEFF2_HUMAN|TENB 2|TENB2|TMEFF2|Tomoregulin|Tomoregulin-2|TPEF|TR 2|TR|TR-2|Transmembrane protein containing epidermal growth factor and follistatin domains|Transmembrane protein TENB2|Transmembrane protein with EGF like and two follistatin like domains 2|Transmembrane protein with EGF-like and two follistatin-like domains
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UIK5
- Q9UIK5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human TMEFF2
- Synthetic peptide of Human TMEFF2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together