Anti-PRMT3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:10000, IHC:1:50 to 1:200
Type:
Antigen: PRMT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77848-457
- 77848-456
- Antigen Symbol:
- PRMT3
- PRMT3
- Antigen Synonyms:
- 2010005E20Rik|2410018A17Rik|AL033309|ANM3_HUMAN|Heterogeneous nuclear ribonucleoprotein methyltransferase 1-like 3|Heterogeneous nuclear ribonucleoprotein methyltransferase like 3|Heterogeneous nuclear ribonucleoprotein methyltransferase like protein 3|Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3|HMT1 hnRNP methyltransferase like 3|HMT1 hnRNP methyltransferase like 3 S. cerevisiae|HMT1-like 3|hnRNP methyltransferase like 3|HRMT1L3|PRMT 3|Protein arginine methyltransferase 3|Protein arginine N methyltransferase 3|Protein arginine N-methyltransferase 3
- 2010005E20Rik|2410018A17Rik|AL033309|ANM3_HUMAN|Heterogeneous nuclear ribonucleoprotein methyltransferase 1-like 3|Heterogeneous nuclear ribonucleoprotein methyltransferase like 3|Heterogeneous nuclear ribonucleoprotein methyltransferase like protein 3|Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3|HMT1 hnRNP methyltransferase like 3|HMT1 hnRNP methyltransferase like 3 S. cerevisiae|HMT1-like 3|hnRNP methyltransferase like 3|HRMT1L3|PRMT 3|Protein arginine methyltransferase 3|Protein arginine N methyltransferase 3|Protein arginine N-methyltransferase 3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60678
- O60678
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human PRMT3
- Synthetic peptide of Human PRMT3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together