Anti-CACNA1A Rabbit Recombinant Polyclonal Antibody
Catalog # 77852-241
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CACNA1A
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:O00555
- Antigen Synonyms:CACH4|CACN3|CACNL1A4|Voltage-dependent P/Q-type calcium channel subunit alpha-1A|Brain calcium channel I|BI|Calcium channel|L type|alpha-1 polypeptide isoform 4|Voltage-gated calcium channel subunit alpha Cav2.1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Voltage-dependent P/Q-type calcium channel subunit alpha-1A protein (1838-2136AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: CACNA1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human