Anti-ARL2BP Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: ARL2BP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77846-109
- 77846-110
- Antigen Symbol:
- ARL2BP
- ARL2BP
- Antigen Synonyms:
- ADP ribosylation factor like 2 binding protein|ADP-ribosylation factor-like protein 2-binding protein|AR2BP_HUMAN|Arf like 2 binding protein BART1|ARF-like 2-binding protein|ARL2 binding protein|ARL2BP protein|BART|BART1|Binder of ARF2 protein 1|Binder of Arl Two|Binder of Arl2|Retinitis pigmentosa 66 (autosomal recessive)|RP66
- ADP ribosylation factor like 2 binding protein|ADP-ribosylation factor-like protein 2-binding protein|AR2BP_HUMAN|Arf like 2 binding protein BART1|ARF-like 2-binding protein|ARL2 binding protein|ARL2BP protein|BART|BART1|Binder of ARF2 protein 1|Binder of Arl Two|Binder of Arl2|Retinitis pigmentosa 66 (autosomal recessive)|RP66
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2Y0
- Q9Y2Y0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human ADP-ribosylation factor-like protein 2-binding protein (1-163AA)
- Recombinant human ADP-ribosylation factor-like protein 2-binding protein (1-163AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together