Anti-DUSP3 Rabbit Recombinant Polyclonal Antibody
Catalog # 77854-425
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DUSP3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P51452
- Antigen Synonyms:Dual specificity phosphatase 3| Dual specificity protein phosphatase 3| Dual specificity protein phosphatase VHR| DUS3_HUMAN| DUSP3| Serine/threonine specific protein phosphatase| Vaccinia H1-related phosphatase| Vaccinia virus phosphatase VH1 related| VHR
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Dual specificity protein phosphatase 3 protein (14-96AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: DUSP3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human