Anti-APCS Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77851-244
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:APCS
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P02743
- Antigen Synonyms:9.5S alpha 1 glycoprotein|9.5S alpha-1-glycoprotein|Amyloid P component serum|MGC88159|Pentaxin related|Pentraxin 2|PTX 2|PTX2|SAMP_HUMAN|SAP|Serum Amyloid P|Serum amyloid P component|Serum amyloid P-component(1-203)
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Serum amyloid P-component protein (20-223AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: APCS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human