Anti-SPN Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:10000, IHC:1:100 to 1:300
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77849-662
- 77849-661
- Antigen Symbol:
- SPN
- SPN
- Antigen Synonyms:
- CD 43|CD43|CD43 antigen|Galactoglycoprotein|GALGP|GPL 115|GPL115|Human gene for sialophorin|Leucocyte sialoglycoprotein|LEUK_HUMAN|Leukocyte large sialoglycoprotein|Leukocyte sialoglycoprotein|Leukosialin|LSN|Ly-48|sialophorin (gpL115, leukosialin, CD43)|Sialophorin|Spn
- CD 43|CD43|CD43 antigen|Galactoglycoprotein|GALGP|GPL 115|GPL115|Human gene for sialophorin|Leucocyte sialoglycoprotein|LEUK_HUMAN|Leukocyte large sialoglycoprotein|Leukocyte sialoglycoprotein|Leukosialin|LSN|Ly-48|sialophorin (gpL115, leukosialin, CD43)|Sialophorin|Spn
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P16150
- P16150
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human SPN
- Fusion protein of Human SPN
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together