Anti-BET1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77851-861
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GAP43
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P17677
- Antigen Synonyms:Axonal membrane protein GAP 43|Axonal membrane protein GAP-43|B 50|Calmodulin binding protein P 57|F1|GAP 43|GAP43|Growth Associated Protein 43|Growth-associated protein 43|Nerve Growth Related Peptide|Nerve growth related peptide GAP43|NEUM_HUMAN|Neural phosphoprotein B 50|Neural phosphoprotein B-50|Neuromodulin|Neuron growth associated protein 43|PP46|Protein F1|QtrA-11580|QtrA-13071
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:QtrA-13071
- Purification:Antigen affinity purified
Specifications
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human