Anti-AP1G1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-208
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:AP1G1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O43747
- Antigen Synonyms:Adapter-related protein complex 1 subunit gamma-1|Adaptor protein complex AP 1 gamma 1 subunit|Adaptor protein complex AP 1 subunit gamma 1|Adaptor protein complex AP-1 subunit gamma-1|Adaptor related protein complex 1 gamma 1 subunit|ADTG|AP 1 complex subunit gamma 1|AP-1 complex subunit gamma-1|AP1G1_HUMAN|CLAPG1|Clathrin assembly protein complex 1 gamma 1 large chain|Clathrin assembly protein complex 1 gamma large chain|Clathrin assembly protein complex 1 gamma-1 large chain|Clathrin associated/assembly/adaptor protein large gamma 1|Gamma1 adaptin|Gamma1-adaptin|Golgi adaptor HA1/AP1 adaptin gamma subunit|Golgi adaptor HA1/AP1 adaptin subunit gamma 1|Golgi adaptor HA1/AP1 adaptin subunit gamma-1|MGC18255
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human AP-1 complex subunit gamma-1 protein (592-686AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: AP1G1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse