Anti-RBP5 Rabbit Polyclonal Antibody
Catalog # 77847-046
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:RBP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Gene ID:P82980
- Antigen Synonyms:Cellular retinol-binding protein III|CRBP III|CRBP-III|CRBP3|CRBPIII|HRBPiso|OTTHUMP00000238163|Putative cellular retinol binding protein CRBP III|RET5_HUMAN|Retinol binding protein|Retinol binding protein 5|cellular|Retinol-binding protein 5
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human CRBP III.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:50 to 1:100, IF: 1:100 to 1:500
Type:
Antigen: RBP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human