Anti-POLI Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: POLI
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77844-808
- 77844-807
- Antigen Symbol:
- POLI
- POLI
- Antigen Synonyms:
- DNA polymerase iota|Eta 2|Eta2|POLI|POLI_HUMAN|Polymerase (DNA directed) iota|RAD 30B|RAD30 homolog B|RAD30, S. cerevisiae, homolog of|RAD30B|RAD3OB
- DNA polymerase iota|Eta 2|Eta2|POLI|POLI_HUMAN|Polymerase (DNA directed) iota|RAD 30B|RAD30 homolog B|RAD30, S. cerevisiae, homolog of|RAD30B|RAD3OB
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UNA4
- Q9UNA4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human DNA polymerase iota protein (458-711AA)
- Recombinant Human DNA polymerase iota protein (458-711AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together