Anti-PAM16 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PAM16
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-655
- 77845-656
- Antigen Symbol:
- PAM16
- PAM16
- Antigen Synonyms:
- CGI-136|MAGMAS|Magmas like protein|Mitochondria associated protein involved in granulocyte macrophage colony stimulating factor signal transduction|Mitochondria-associated granulocyte macrophage CSF-signaling molecule|Mitochondrial import inner membrane translocase subunit TIM16|Presequence translocase-associated motor 16 homolog (S. cerevisiae)|Presequence translocated-associated motor subunit TIM16|TIM16_HUMAN|TIMM16
- CGI-136|MAGMAS|Magmas like protein|Mitochondria associated protein involved in granulocyte macrophage colony stimulating factor signal transduction|Mitochondria-associated granulocyte macrophage CSF-signaling molecule|Mitochondrial import inner membrane translocase subunit TIM16|Presequence translocase-associated motor 16 homolog (S. cerevisiae)|Presequence translocated-associated motor subunit TIM16|TIM16_HUMAN|TIMM16
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3D7
- Q9Y3D7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human mitochondrial import inner membrane translocase subunit TIM16 protein (1-125AA)
- Recombinant human mitochondrial import inner membrane translocase subunit TIM16 protein (1-125AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together