Anti-RAB1A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
77852-019
New Product
- Antigen Symbol:RAB1A
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P62820
- Antigen Synonyms:GTP binding protein RAB 1A|mKIAA3012|RAB 1|Rab 1A|RAB1|RAB1, member RAS oncogene family|Rab1A|RAB1A member RAS oncogene family|RAB1A_HUMAN|Ras related protein Rab 1A|Ras-associated protein RAB1|Ras-related protein Rab-1A|YPT1|YPT1 related protein|YPT1-related protein
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Ras-related protein Rab-1A protein (2-205AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: RAB1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human