Anti-SENP5 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000
Type:
Antigen: SENP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77849-905
- 77849-904
- Antigen Symbol:
- SENP5
- SENP5
- Antigen Synonyms:
- FKSG45|FLJ42398|MGC27076|Protease FKSG45|SENP5|SENP5_HUMAN|Sentrin specific protease SENP5|Sentrin-specific protease 5|Sentrin/SUMO specific protease SENP5|Sentrin/SUMO-specific protease SENP5|SUMO specific protease SENP5|SUMO1/sentrin specific peptidase 5|SUMO1/sentrin specific protease 5
- FKSG45|FLJ42398|MGC27076|Protease FKSG45|SENP5|SENP5_HUMAN|Sentrin specific protease SENP5|Sentrin-specific protease 5|Sentrin/SUMO specific protease SENP5|Sentrin/SUMO-specific protease SENP5|SUMO specific protease SENP5|SUMO1/sentrin specific peptidase 5|SUMO1/sentrin specific protease 5
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96HI0
- Q96HI0
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human SENP5
- Fusion protein of Human SENP5
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together