Anti-KPNA6 Rabbit Recombinant Polyclonal Antibody
Catalog # 77858-706
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:KPNA6
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Form:Liquid
- Gene ID:O60684
- Antigen Synonyms:FLJ11249|IMA7_HUMAN|Importin alpha 7 subunit|Importin alpha S2|Importin subunit alpha 7|Importin subunit alpha-7|IPOA 7|IPOA7|Karyopherin alpha 6|Karyopherin alpha 6 (importin alpha 7)|Karyopherin subunit alpha 6|Karyopherin subunit alpha-6|KPNA 6|KPNA 7|KPNA7|MGC17918|OTTHUMP00000004532
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Importin subunit alpha-7 protein (277-536AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: KPNA6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human