Anti-ATP6AP2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-630
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ATP6AP2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:O75787
- Antigen Synonyms:APT6M8 9|APT6M8-9|ATP6AP2|ATP6IP2|ATP6M8-9|ATPase H(+)-transporting lysosomal accessory protein 2|ATPase H(+)-transporting lysosomal-interacting protein 2|ATPase H+ transporting lysosomal accessory protein 2|ATPase H+ transporting lysosomal interacting protein 2|ATPase H+ transporting lysosomal vacuolar proton pump membrane sector associated protein M8 9|ATPase membrane sector associated protein M8 9|ATPase|H+ transporting|lysosomal (vacuolar proton pump) membrane sector associated protein M8 9|CAPER|ELDF10|Embryonic liver differentiation factor 10|ER localized type I transmembrane adaptor|ER-localized type I transmembrane adaptor|HT028|M8 9|M8-9|MGC99577|MRXE|MSTP009|N14F|Renin receptor|Renin/prorenin receptor|RENR_HUMAN|V ATPase M8 9 subunit|V ATPase M8.9 subunit|V-ATPase M8.9 subunit|Vacuolar ATP synthase membrane sector associated protein M8 9|Vacuolar ATP synthase membrane sector-associated protein M8-9|vacuolar proton ATP synthase membrane sector associated protein M8 9|XMRE
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Renin receptor protein (251-350AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ATP6AP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse