Anti-CD96 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77852-778
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CD96
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P40200
- Antigen Synonyms:CD96 molecule|Cell surface antigen DKFZp667E2122|MGC22596|T cell activated increased late expression protein|T cell activation|increased late expression|T cell surface protein tactile precursor|T cell-activated increased late expression protein|T-cell surface protein tactile|TACT_HUMAN|Tactile
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human T-cell surface protein tactile protein (306-395AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CD96
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human