Anti-ANXA6 Rabbit Recombinant Polyclonal Antibody
77855-091
New Product
- Antigen Symbol:ANXA6
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P08133
- Antigen Synonyms:67 kDa calelectrin|Annexin A6|Annexin VI|Annexin VI p68|Annexin-6|AnnexinA6|AnnexinVI|ANX 6|ANX A6|ANX6|ANXA 6|ANXA6_HUMAN|Calcium binding protein p68|Calelectrin|Calphobindin II|Calphobindin-II|CalphobindinII|CBP 68|CBP68|Chromobindin 20|Chromobindin-20|Chromobindin20|CPB II|CPB-II|CPBII|Lipocortin VI|LipocortinVI|p68|p70|Protein III|ProteinIII
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Annexin A6 protein (2-245AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: ANXA6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat