Anti-CHKB Rabbit Recombinant Polyclonal Antibody (Biotin)
Catalog # 77853-072
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CHKB
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:Q9Y259
- Antigen Synonyms:CHETK|Chetk-beta|Chkb|Chkb choline kinase beta|Chkb protein|CHKB_HUMAN|CHKL|Choline kinase beta|Choline kinase-like|Choline kinase-like protein|Choline/ethanolamine kinase|choline/ethanolamine kinase beta|beta|CK|CK/EK|CK/EK-beta|CKB|CKEKB|EK|EKB|Ethanolamine kinase|Ethanolamine kinase beta|MDCMC
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Choline/ethanolamine kinase protein (2-99AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CHKB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human