Anti-TMCO1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TMCO1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-503
- 77844-502
- Antigen Symbol:
- TMCO1
- TMCO1
- Antigen Synonyms:
- 1190006A08Rik|4930403O06Rik|AA109065|AU022572|Calcium load activated calcium channel|CFSMR|CLAC channel|ESTM39|HP10122|Meg 2 like protein|PCIA3|PNAS 10|PNAS 136|Putative membrane protein|RP11-466F5.7|TMCC4|TMCO1_HUMAN|Transmembrane and coiled coil domains 1|Transmembrane and coiled coil domains 4|Transmembrane and coiled-coil domain-containing protein 1|Transmembrane and coiled-coil domains protein 4|Xenogeneic cross-immune protein PCIA3
- 1190006A08Rik|4930403O06Rik|AA109065|AU022572|Calcium load activated calcium channel|CFSMR|CLAC channel|ESTM39|HP10122|Meg 2 like protein|PCIA3|PNAS 10|PNAS 136|Putative membrane protein|RP11-466F5.7|TMCC4|TMCO1_HUMAN|Transmembrane and coiled coil domains 1|Transmembrane and coiled coil domains 4|Transmembrane and coiled-coil domain-containing protein 1|Transmembrane and coiled-coil domains protein 4|Xenogeneic cross-immune protein PCIA3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UM00
- Q9UM00
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Calcium load-activated calcium channel protein (43-79AA)
- Recombinant human Calcium load-activated calcium channel protein (43-79AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together