Anti-DPP6 Rabbit Recombinant polyclonal Antibody (Biotinylated)
77854-337
New Product
- Antigen Symbol:DPP6
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P42658
- Antigen Synonyms:DPP6Dipeptidyl aminopeptidase-like protein 6|DPPX|Dipeptidyl aminopeptidase-related protein|Dipeptidyl peptidase 6|Dipeptidyl peptidase IV-like protein|Dipeptidyl peptidase VI|DPP VI
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Dipeptidyl aminopeptidase-like protein 6 protein (117-859AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
0
Type:
Antigen: DPP6
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human