Anti-CA9 HEK293F cells Recombinant Monoclonal Antibody [clone: 4F12]
Catalog # 77850-374
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CA9
- Clonality:Recombinant Monoclonal
- Clone:4F12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293F cells
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:Q16790
- Antigen Synonyms:Carbonic anhydrase 9, Carbonate dehydratase IX, Carbonic anhydrase IX, CA-IX, CAIX, Membrane antigen MN, P54/58N, Renal cell carcinoma-associated antigen G250, RCC-associated antigen G250, pMW1, CA9, G250, MN
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:A synthesized peptide derived from Human CA9
- Purification:Affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:50 to 1:200
Type:
Antigen: CA9
Clonality: Recombinant Monoclonal
Clone: 4F12
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: IgG
Reactivity: Human