Anti-EFNA2 Rabbit Polyclonal Antibody
77848-589
New Product
- Antigen Symbol:EFNA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O43921
- Antigen Synonyms:EFNA 2 |efna2|EFNA2_HUMAN|ELF1|Eph related receptor tyrosine kinase ligand 6|EPH-related receptor tyrosine kinase ligand 6|Ephrin-A2|EphrinA2|EPLG6|HEK7 L|HEK7 ligand|HEK7-L|HEK7L|LERK 6|LERK-6|LERK6|Ligand of eph related kinase 6
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Internal of Human EFNA2.
- Purification:Antigen affinity purified
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human