Anti-CD99 HEK293F cells Recombinant Monoclonal Antibody [clone: 9A5]
Catalog # 77850-182
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CD99
- Clonality:Recombinant Monoclonal
- Clone:9A5
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:HEK293F cells
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P14209
- Antigen Synonyms:CD99 antigen|12E7|E2 antigen|Protein MIC2|T-cell surface glycoprotein E2|MIC2|MIC2X|MIC2Y
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide
- Purification:Affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:50 to 1:500
Type:
Antigen: CD99
Clonality: Recombinant Monoclonal
Clone: 9A5
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: IgG
Reactivity: Human