Anti-CDC25B Rabbit Recombinant Polyclonal Antibody
Catalog # 77852-784
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CDC25B
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P30305
- Antigen Synonyms:AI604853|Cdc 25B|Cdc25b|CDC25HU2|Cdc25m2|Cell division cycle 25 homolog B|Cell division cycle 25B|Cell division cycle 25B isoform 1|Cell division cycle 25B isoform 2|Cell division cycle 25B isoform 3|Cell division cycle 25B isoform 4|Cell division cycle 25B isoform 5|Dual specificity phosphatase Cdc25B|EC 3.1.3.48|M phase inducer phosphatase 2|M-phase inducer phosphatase 2|MPIP2_HUMAN
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human M-phase inducer phosphatase 2 protein (1-195AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: CDC25B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human