Human Recombinant FGF19 (from HEK293 Cells)
Supplier: Antibodies.com
Certificates
About this item
Partial length Human FGF19, expressed in HEK293 cells.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:Lyophilized: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# O95750
- Reconstitution Instructions:Reconstitute with distilled sterile water
- Carrier-Free:N
- Protein Synonyms:FGF-19|Fibroblast growth factor 19
- Protein/Peptide Name:FGF19
- Purity:>85% by SDS-PAGE and Coomassie blue staining
- Molecular Weight:The protein has a predicted molecular mass of 22.3 kDa after removal of the signal peptide. The apparent molecular mass of FGF19-His is approximately 15 - 25 kDa due to glycosylation.
- Sequence:FGF19 (Leu25-Lys216) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% Trehalose is added as a protectant before lyophilization.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Tested Applications:SDS-PAGE
Specifications
Frequently Bought Together