Anti-STK24 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77851-748
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:STK24
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q9Y6E0
- Antigen Synonyms:epididymis secretory protein Li 95|HEL-S-95|Mammalian STE20 like protein kinase 3|Mammalian STE20-like protein kinase 3|Mammalian STE20-like protein kinase 3 C-terminal|Mammalian STE20-like protein kinase 3 N-terminal|Mammalian sterile 20-like 3|MST-3|MST3/C|MST3/N|MST3B|serine threonine kinase 24|Serine/threonine kinase 24 (Ste20|yeast homolog)|Serine/threonine-protein kinase 24 12 kDa subunit|Serine/threonine-protein kinase 24|STE20 homolog yeast|STE20 like kinase MST3|STE20-like kinase MST3|STK24_HUMAN|STK3
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Serine/threonine-protein kinase 24 protein (1-431AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: STK24
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human