Anti-KCNK9 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KCNK9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-360
- 77843-359
- Antigen Symbol:
- KCNK9
- KCNK9
- Antigen Synonyms:
- TASK3Potassium channel subfamily K member 9|Acid-sensitive potassium channel protein TASK-3|TWIK-related acid-sensitive K(+) channel 3|Two pore potassium channel KT3.2|Two pore K(+) channel KT3.2
- TASK3Potassium channel subfamily K member 9|Acid-sensitive potassium channel protein TASK-3|TWIK-related acid-sensitive K(+) channel 3|Two pore potassium channel KT3.2|Two pore K(+) channel KT3.2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NPC2
- Q9NPC2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Potassium channel subfamily K member 9 protein (250-362AA)
- Recombinant Human Potassium channel subfamily K member 9 protein (250-362AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together