Order Entry
United States
ContactUsLinkComponent
Anti-MT-ND6 Rabbit Polyclonal Antibody
Anti-MT-ND6 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MT-ND6 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77848-889
  • 77848-890
  • Antigen Symbol:
  • MT-ND6
  • MT-ND6
  • Antigen Synonyms:
  • MTND6|NADH6|ND6|NADH-ubiquinone oxidoreductase chain 6|EC 7.1.1.2|NADH dehydrogenase subunit 6
  • MTND6|NADH6|ND6|NADH-ubiquinone oxidoreductase chain 6|EC 7.1.1.2|NADH dehydrogenase subunit 6
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P03923
  • P03923
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide of Human MT-ND6
  • Synthetic peptide of Human MT-ND6
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together