Anti-PLCL1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: PLCL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77848-591
- 77848-590
- Antigen Symbol:
- PLCL1
- PLCL1
- Antigen Synonyms:
- Inactive phospholipase C-like protein 1| Inactive protein 1| Phospholipase C deleted in lung carcinoma| Phospholipase C epsilon| Phospholipase C like 1| Phospholipase C related but catalytically inactive protein| Phospholipase C related, but catalytically inactive protein| Phospholipase C-deleted in lung carcinoma| Phospholipase C-related but catalytically inactive protein| Phospholipase C-related catalytically| PLC-L1| PLCE| PLCL| PLCL1_HUMAN| PLDL1| PRIP
- Inactive phospholipase C-like protein 1| Inactive protein 1| Phospholipase C deleted in lung carcinoma| Phospholipase C epsilon| Phospholipase C like 1| Phospholipase C related but catalytically inactive protein| Phospholipase C related, but catalytically inactive protein| Phospholipase C-deleted in lung carcinoma| Phospholipase C-related but catalytically inactive protein| Phospholipase C-related catalytically| PLC-L1| PLCE| PLCL| PLCL1_HUMAN| PLDL1| PRIP
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15111
- Q15111
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human PLCL1
- Synthetic peptide of Human PLCL1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together