PKCε Peptide Substrate
Catalog # 80054-014
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Size:500 μg
- Protein Synonyms:Protein Kinase C ε Peptide Substrate|Peptide-ε|Protein Kinase Cε Peptide Substrate|ERMRPRKRQGSVRRRV
- Protein/Peptide Name:PKCε Peptide Substrate
- Purity:≥95% (HPLC)
Specifications
About this item
PKCε Peptide Substrate, Pseudosubstrate region of the ε-isotype of protein kinase C (PKCε), with an alanine to serine substitution.
Pseudosubstrate region of the ε-isotype of protein kinase C (PKCε) with an alanine to serine substitution. PKCε exhibits an apparent specificity for this peptide (Kₘ = 68 µM).