Anti-ITGB7 Rabbit Recombinant Polyclonal Antibody
Catalog # 77858-329
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ITGB7
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Antigen Synonyms:Gut homing receptor beta subunit|Integrin beta 7 subunit|Integrin beta-7|Integrin beta-P|Integrin|beta 7|ITB7|ITB7_HUMAN|Itgb7|Ly69|M290 IEL antigen
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Integrin beta-7 protein (1-140AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ITGB7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human