Anti-ATE1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-515
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ATE1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O95260
- Antigen Synonyms:AI225793| Arginine tRNA protein transferase 1| Arginine-tRNA--protein transferase 1| Arginyl-tRNA--protein transferase 1| Arginyl-tRNA-protein transferase| Arginyltransferase 1| Ate1| ATE1_HUMAN| AW547406| EC 2.3.2.8| MGC26724| OTTHUMP00000020637| OTTHUMP00000058665| R transferase 1| R-transferase 1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Arginyl-tRNA--protein transferase 1 protein (105-253AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: ATE1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human