Anti-DCTN4 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: DCTN4
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77845-076
- 77845-077
- Antigen Symbol:
- DCTN4
- DCTN4
- Antigen Synonyms:
- DCTN 4|DCTN4_HUMAN|Dynactin 4 (p62)|Dynactin 4|Dynactin p62 subunit|Dynactin subunit 4|Dynactin subunit p62|Dynactin4|p62
- DCTN 4|DCTN4_HUMAN|Dynactin 4 (p62)|Dynactin 4|Dynactin p62 subunit|Dynactin subunit 4|Dynactin subunit p62|Dynactin4|p62
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UJW0
- Q9UJW0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Dynactin subunit 4 protein (2-460AA)
- Recombinant Human Dynactin subunit 4 protein (2-460AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together