Streptavidin AP Conjugate, AP (Alkaline Phosphatase)
Catalog # EM69219-3
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Conjugation:AP (Alkaline Phosphatase)
- Size:250 μl
- Storage Conditions:Safe to freeze, avoid repeated freeze/thaw cycles
- Protein/Peptide Name:Streptavidin AP Conjugate