Anti-FABP4 HEK293F cells Recombinant Monoclonal Antibody [clone: 1H4]
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FABP4
Clonality: Recombinant Monoclonal
Clone: 1H4
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77850-206
- 77850-207
- Antigen Symbol:
- FABP4
- FABP4
- Antigen Synonyms:
- Fatty acid-binding protein, adipocyte, Adipocyte lipid-binding protein, ALBP, Adipocyte-type fatty acid-binding protein, A-FABP, AFABP, Fatty acid-binding protein 4, FABP4
- Fatty acid-binding protein, adipocyte, Adipocyte lipid-binding protein, ALBP, Adipocyte-type fatty acid-binding protein, A-FABP, AFABP, Fatty acid-binding protein 4, FABP4
- Clonality:
- Recombinant Monoclonal
- Recombinant Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 1H4
- 1H4
- Reactivity:
- Human
- Human
- Host:
- HEK293F cells
- HEK293F cells
- Gene ID:
- P15090
- P15090
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- A synthesized peptide derived from Human FABP4
- A synthesized peptide derived from Human FABP4
- Form:
- Liquid
- Liquid
- Purification:
- Affinity purified
- Affinity purified
- Storage Buffer:
- PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together