Anti-AADAC Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77850-494
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:AADAC
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P22760
- Antigen Synonyms:AAAD_HUMAN|Aada|Arylacetamide deacetylase (esterase)|Arylacetamide deacetylase|CES5A1|DAC
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Arylacetamide deacetylase protein (24-399AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: AADAC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human