Anti-FOXO4 (phospho-Thr451) Rabbit Polyclonal Antibody
77848-146
New Product
- Antigen Symbol:MFF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9GZY8
- Antigen Synonyms:C2orf33 |Chromosome 2 open reading frame 33|DKFZp666J168|GL004 |Mff|MFF_HUMAN|MGC110913|Mitochondrial fission factor|OTTHUMP00000164235
- Modification:phospho-Thr451
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:OTTHUMP00000164235
- Purification:Antigen affinity purified
Recommended dilution: WB: 1:500 to 1:1000
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human