Anti-CHKA Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77853-068
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CHKA
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P35790
- Antigen Synonyms:CHK|CKI|Choline kinase alpha|CK|EC 2.7.1.32|CHETK-alpha|Ethanolamine kinase|EK|EC 2.7.1.82
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Choline kinase alpha protein (79-173AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CHKA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human