Anti-ABCB4 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77850-514
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ABCB4
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P21439
- Antigen Synonyms:ABC 21|ABC B4|ABC21|ABCB 4|Abcb4|ABCB4 protein|ATP binding cassette sub family B MDR/TAP member 4|ATP binding cassette sub family B member 4|ATP-binding cassette sub-family B member 4|GBD1|ICP3|MDR 3|MDR2|MDR2/3|MDR3|MDR3 P glycoprotein|MDR3 P gp|MDR3_HUMAN|Multidrug resistance protein 3|Multiple drug resistance 3|P glycoprotein 3|P-glycoprotein 3|PFIC 3|PFIC3|PGY 3|PGY3
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Phosphatidylcholine translocator ABCB4 protein (530-693AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ABCB4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human