Anti-HOXD9 Rabbit Recombinant Polyclonal Antibody
Catalog # 77857-616
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HOXD9
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P28356
- Antigen Synonyms:homeobox D9|Homeobox protein Hox D9|Homeobox protein Hox-4C|Homeobox protein Hox-5.2|Homeobox protein Hox-D9|Hox 4.3|Hox 5.2|HOX4|HOX4C|HXD9_HUMAN|RP23 313J15.10
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Homeobox protein Hox-D9 protein (38-276AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:2000 to 1:10000, IHC: 1:20 to 1:200
Type:
Antigen: HOXD9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse