Anti-ADAMTS7 Rabbit Recombinant Polyclonal Antibody
Catalog # 77844-612
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ADAMTS7
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9UKP4
- Antigen Synonyms:A disintegrin and metalloprotease with thrombospondin motifs 7 preproprotein|A disintegrin and metalloproteinase with thrombospondin motifs 7|A disintegrin like and metalloprotease (reprolysin type) with thrombospondin type 1 motif 7|A disintegrin like and metalloprotease with thrombospondin type 1 motif 7|ADAM metallopeptidase with thrombospondin type 1 motif 7|ADAM metallopeptidase with thrombospondin type 1 motif 7 preproprotein|ADAM TS 7|ADAM TS7|ADAM-TS 7|ADAM-TS7|ADAMTS 7|ADAMTS-7|Adamts7|ATS7_HUMAN|COMPase|DKFZp434H204
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human A disintegrin and metalloproteinase with thrombospondin motifs 7 protein (242-593AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: ADAMTS7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse