Anti-BLVRB Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-902
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:BLVRB
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Form:Liquid
- Gene ID:P30043
- Antigen Synonyms:Biliverdin IX beta reductase|biliverdin reductase B (flavin reductase (NADPH))|Biliverdin reductase B|Biliverdin-IX beta-reductase|BLVRB|BLVRB_HUMAN|BVR B|BVR-B|BVRB|epididymis secretory protein Li 10|Flavin reductase (NADPH)|Flavin reductase|FLR|FR|GHBP|Green heme binding protein|Green heme-binding protein|HEL-S-10|Methemoglobin reductase|MGC117413|NADPH dependent diaphorase|NADPH flavin reductase|NADPH-dependent diaphorase|NADPH-flavin reductase|SDR43U1|short chain dehydrogenase/reductase family 43U|member 1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Flavin reductase (NADPH) protein (2-206AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000, IHC:1:200 to 1:500, IF:1:50 to 1:200
Type:
Antigen: BLVRB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human