Anti-CDK12 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: CDK12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77850-102
- 77850-101
- Antigen Symbol:
- CDK12
- CDK12
- Antigen Synonyms:
- arginine/serine-rich|Cdc2 related kinase arginine/serine rich|CDC2 related protein kinase 7|Cdc2-related kinase|CDC2-related protein kinase 7|CDK12_HUMAN|Cell division cycle 2 related protein kinase 7|Cell division cycle 2-related protein kinase 7|Cell division protein kinase 12|CRK7|CRKR|CrkRS|Cyclin-dependent kinase 12|EC 2.7.11.22|hkiaa0904|Pksc
- arginine/serine-rich|Cdc2 related kinase arginine/serine rich|CDC2 related protein kinase 7|Cdc2-related kinase|CDC2-related protein kinase 7|CDK12_HUMAN|Cell division cycle 2 related protein kinase 7|Cell division cycle 2-related protein kinase 7|Cell division protein kinase 12|CRK7|CRKR|CrkRS|Cyclin-dependent kinase 12|EC 2.7.11.22|hkiaa0904|Pksc
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NYV4
- Q9NYV4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human CDK12
- Synthetic peptide of Human CDK12
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together