Anti-H3F3A HEK293F cells Recombinant Monoclonal Antibody [clone: 4H2]
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:50 to 1:500, IF:1:30 to 1:200
Type:
Antigen: H3F3A
Clonality: Recombinant Monoclonal
Clone: 4H2
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77850-238
- 77850-237
- Antigen Symbol:
- H3F3A
- H3F3A
- Antigen Synonyms:
- Histone H3.3, H3F3A, H3.3A, H3F3, PP781, AND, H3F3B, H3.3B
- Histone H3.3, H3F3A, H3.3A, H3F3, PP781, AND, H3F3B, H3.3B
- Clonality:
- Recombinant Monoclonal
- Recombinant Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 4H2
- 4H2
- Reactivity:
- Human
- Human
- Host:
- HEK293F cells
- HEK293F cells
- Gene ID:
- P84243
- P84243
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Synthesized peptide
- Synthesized peptide
- Form:
- Liquid
- Liquid
- Purification:
- Affinity purified
- Affinity purified
- Storage Buffer:
- PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together