Anti-AMT Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-119
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:AMT
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P48728
- Antigen Synonyms:Aminomethyltransferase (glycine cleavage system protein T)|Aminomethyltransferase|AMT|GCE|GCST|GCST_HUMAN|GCVT|Glycine cleavage system T protein|mitochondrial|NKH
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Aminomethyltransferase, mitochondrial protein (180-260AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000
Type:
Antigen: AMT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse